Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karl Fischer Reagent Solution A, for pyridine-based volumetric KF titration (pyridine, sulfur dioxide, methanol), Solstice

Catalog No. 60046639 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-639 6 x 1 L
60-008-81 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-639 Supplier Solstice Supplier No. 361166X1L
Add to Cart
Add to Cart

for pyridine-based volumetric Karl Fischer titration (pyridine, sulfur dioxide, methanol)

  • Solutions are prepared with raw materials which meet or exceed requirements of the American Chemical Society
  • Where no ACS specifications exist, chemicals of highest purity available are used
  • All aqueous solutions are prepared using high purity, deionized, water which meets requirements for ASTM type 1 Reagent Grade Water
  • Exacting production and quality control records are maintained providing traceability of raw materials
  • All solutions are guaranteed to meet label specifications
  • All solution labels meet or exceed all ISO, DOT and OSHA regulations
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Karl Fischer Reagent Solution A
Name Note for pyridine-based volumetric KF titration (pyridine, sulfur dioxide, methanol)
Boiling Point >60°C
CAS Min % ≥10.0000%, ≥25.0000%
CAS Max % <20.0000%, <50.0000%
CAS 7446-09-5, 110-86-1
Density ∼1.01 g/mL
Quantity 6 x 1 L
Flash Point 16°C
Packaging Glass Bottle
UN Number UN1992
Grade Karl Fischer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.