Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER REAGENT, 5 mg/ml, STABILIZED, PYRIDINE-BASED | 6 X 1 L | GFS CHEMICALS, INC. Item #: 99904 | Bulk #: 1620
SDP
Catalog No. NC0123606
Encompass_Preferred
WATERMARK KARL FISCHER REAGENT, 5 mg/ml, STABILIZED, PYRIDINE-BASED | For volumetric titrations. ; Specifications : Suitability, Titer strength (mg/mL) 5.0 - 6.0 mg/mL
Catalog No. NC0123606 Supplier GFS Chemicals Inc Supplier No. 99904
WATERMARK KARL FISCHER REAGENT, 5 mg/ml, STABILIZED, PYRIDINE-BASED | For volumetric titrations. ; Specifications : Suitability, Titer strength (mg/mL) 5.0 - 6.0 mg/mL