Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karl Fischer Solvent KT, solvent for volumetric one- and two-component titration, for aldehydes and ketones, Thermo Scientific™

Catalog No. p-7052336
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA47219K2 1 L
AA47219Y0 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA47219K2 Supplier Thermo Scientific Chemicals Supplier No. 47219K2

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Karl Fischer Solvent KT
Name Note solvent for volumetric one- and two-component titration, for aldehydes and ketones
Boiling Point 120°C
Color Colorless to Yellow
Density 0.940g/mL
Physical Form Liquid
Quantity 1 L
UN Number UN2924
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.