Learn More
Karl Fischer Solvent, Reagents
Specifications
Specifications
| Chemical Name or Material | Karl Fischer Solvent |
| CAS | 67-56-1, 110-86-1, 110-86-1, 67-56-1 |
| Color | Colorless to Slight Yellow |
| Physical Form | Liquid |
| Quantity | 4 L |
| Packaging | Safety Coated Glass |
| Specific Gravity | 0.936 |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.