Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karl Fischer Volumetric Reagent, MilliporeSigma™

Catalog No. MKX00106 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MKX00106 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MKX00106 Supplier MilliporeSigma Supplier No. KX00106

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy
  • Reagent for Volumetric Karl Fischer Titrations.
  • For titrimetric moisture determination. Titer ≈ 5mg H&subscript_2;O/mL.
  • Contains pyridine and 2-methoxyethanol.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Karl Fischer Volumetric Reagent
Quantity 1 L
Packaging Poly-coated Amber Glass
UN Number 1993
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.