Learn More
Electron Microscopy Sciences Karnovsky's Fixative Kit
SDP
Catalog No. 50980497
Encompass_Preferred
Ready To mix. Kit consists of: 2x10 ml ampoules 16% Formaldehyde1x10 ml ampoules 50% Glutaraldehyde1x50 ml 0.2M Sodium Phosphate Buffer Kit consists of: 2 x 10ml ampoules 16% Formaldehyde, 1 x 10ml ampoules 50% Glutaraldehyde, 1 x 50ml 0.2M Sodium Phosphate Buffer.
Catalog No. 50-980-497 Supplier Electron Microscopy Sciences Supplier No. 15730
Ready To mix. Kit consists of: 2x10 ml ampoules 16% Formaldehyde1x10 ml ampoules 50% Glutaraldehyde1x50 ml 0.2M Sodium Phosphate Buffer Kit consists of: 2 x 10ml ampoules 16% Formaldehyde, 1 x 10ml ampoules 50% Glutaraldehyde, 1 x 50ml 0.2M Sodium Phosphate Buffer.