Learn More
Electron Microscopy Sciences Paraformaldehyde-Glutaraldehyde Karnovsky's Fixative, 1 kit
SDP
Catalog No. 50980496
Encompass_Preferred
Ready To mix. Kit consists of: 2x10 ml ampoules 16% Formaldehyde1x10 ml ampoules 50% Glutaraldehyde1x50 ml 0.2M Sodium Phosphate Buffer
Catalog No. 50-980-496 Supplier Electron Microscopy Sciences Supplier No. 15720
Ready To mix. Kit consists of: 2x10 ml ampoules 16% Formaldehyde1x10 ml ampoules 50% Glutaraldehyde1x50 ml 0.2M Sodium Phosphate Buffer