Learn More
Electron Microscopy Sciences Karnovsky Fixative Reagent Grade 1 LT
SDP
Supplier:  Electron Microscopy Sciences 1573110
Encompass_Preferred
This fixative contains 3% glutaraldehyde and 2% formaldehyde in 0.1M phosphate buffer, pH 7.4.
pH: 7.4
Catalog No. NC1484722