Learn More
MSC Karp Access Door, Steel, 24 x 12 in, 1 ea
Catalog No. 5000782639
Encompass
The DSC-214M is a superior access door for new installations or remodeling. Its exposed flange adds rigidity to the unit and perfectly conceals the rough wall opening. Flexible anchor straps can be provided for masonry installation.
  • Designed for walls or ceilings
  • Features two latches
  • Latch is a stainless steel screwdriver cam type
  • Frame constructed from 16 gauge steel
  • Door constructed from 14 gauge steel
  • Trim is 1 inch wide, one-piece construction
Catalog No. 50-007-82639 Supplier MSC Supplier No. 86762069
The DSC-214M is a superior access door for new installations or remodeling. Its exposed flange adds rigidity to the unit and perfectly conceals the rough wall opening. Flexible anchor straps can be provided for masonry installation.
  • Designed for walls or ceilings
  • Features two latches
  • Latch is a stainless steel screwdriver cam type
  • Frame constructed from 16 gauge steel
  • Door constructed from 14 gauge steel
  • Trim is 1 inch wide, one-piece construction