Learn More
Cayman Chemical KarrIkInolID 3ethyl STR 100ug
SDP
Catalog No. 502992241
Encompass_Preferred
A synthetic intermediate; has been used in the synthesis of butanolide seed germination stimulants
Catalog No. 50-299-2241 Supplier Cayman Chemical Supplier No. 31675100 UG
A synthetic intermediate; has been used in the synthesis of butanolide seed germination stimulants