Learn More
Aviva Systems Biology Corp KARS Antibody - C-terminal region (ARP40589_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40589_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KARS. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human KARS. Alternate names: KRS, KARS2, CMTRIBApplicationWB, IHC. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57537