Learn More
TARGETMOL CHEMICALS INC KARTOGENIN 50MG
SDP
Catalog No. 502581101
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Kartogenin (KGN) is an activator of the smad4/smad5 pathway and promotes the selective differentiation of multipotent mesenchymal stem cells into chondrocytes. purity: 96%
Catalog No. 50-258-1101 Supplier TARGETMOL CHEMICALS INC Supplier No. T174850MG
Add to Cart
Add to Cart
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Kartogenin (KGN) is an activator of the smad4/smad5 pathway and promotes the selective differentiation of multipotent mesenchymal stem cells into chondrocytes. purity: 96%