Learn More
Abcam Kartogenin, stem cell differentiation, 25MG
Catalog No. 502706830
Encompass_Preferred
MW 317.3 Da, Purity >98%. Promotes differentiation of human mesenchymal stems cells into chondrocytes (EC₅₀ = 100 nM). Partially supresses release of nitric oxide. Active in vivo and cell permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6830 Supplier Abcam Supplier No. AB14118025MG
MW 317.3 Da, Purity >98%. Promotes differentiation of human mesenchymal stems cells into chondrocytes (EC₅₀ = 100 nM). Partially supresses release of nitric oxide. Active in vivo and cell permeable.
The product is subject to the following: Abcam Restricted Use Statement