Learn More
New England Biolabs, Inc. KasI – 1250 units
SDP
Catalog No. 50812218
Encompass
A restriction endonuclease that recognizes the sequence G^GCGC_C.
Catalog No. 50-812-218 Supplier New England Biolabs, Inc. Supplier No. R0544L
A restriction endonuclease that recognizes the sequence G^GCGC_C.