Learn More
New England Biolabs, Inc. KasI – 250 units
SDP
Catalog No. 50812219
Encompass
A restriction endonuclease that recognizes the sequence G^GCGC_C.
Catalog No. 50-812-219 Supplier New England Biolabs, Inc. Supplier No. R0544S
A restriction endonuclease that recognizes the sequence G^GCGC_C.