Learn More
Next Advance Inc KASIL FORMAMIDE FRIT KIT
SDP
Catalog No. NC0263146
Encompass
Kit to create frits in capillaries. Includes formamide, Kasil-1, Kasil-1624 and a cleaving tool.
Catalog No. NC0263146 Supplier Next Advance Inc Supplier No. FRITKIT
Kit to create frits in capillaries. Includes formamide, Kasil-1, Kasil-1624 and a cleaving tool.