Learn More
Cayman Chemical Kassinin (frog) (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502919265
Encompass_Preferred
A tachykinin neuropeptide; agonist of NK2 and NK3 receptors; selective for the NK2 and NK3 over NK1; increases the short-circuit current in isolated frog skin at 1 µM; induces contractions in isolated guinea pig ileum and isolated rabbit jejunum; decreases blood pressure in anesthetized guinea pigs (ED50 = 1,832 pmol/kg)
Catalog No. 50-291-9265 Supplier Cayman Chemical Supplier No. 3972710 MG
A tachykinin neuropeptide; agonist of NK2 and NK3 receptors; selective for the NK2 and NK3 over NK1; increases the short-circuit current in isolated frog skin at 1 µM; induces contractions in isolated guinea pig ileum and isolated rabbit jejunum; decreases blood pressure in anesthetized guinea pigs (ED50 = 1,832 pmol/kg)