Learn More
Medchemexpress LLC Kasugamycin | 6980-18-3 | 99.8% | 1 G
SDP
Catalog No. 5000409903
Encompass_Preferred
Kasugamycin (Ksg) is an antibiotic that binds to both the 30S and 70S ribosome, but not isolated 50S subunits. It mimics mRNA nucleotides to destabilize tRNA binding and inhibit canonical translation initiation.
  • Binds to 30S and 70S ribosomes
  • Inhibits canonical translation initiation
  • Destabilizes tRNA binding
  • Purity of 99.82%
  • Suitable for research use
Catalog No. 50-004-09903 Supplier Medchemexpress LLC Supplier No. HYB18641G
Kasugamycin (Ksg) is an antibiotic that binds to both the 30S and 70S ribosome, but not isolated 50S subunits. It mimics mRNA nucleotides to destabilize tRNA binding and inhibit canonical translation initiation.
  • Binds to 30S and 70S ribosomes
  • Inhibits canonical translation initiation
  • Destabilizes tRNA binding
  • Purity of 99.82%
  • Suitable for research use