Learn More
Medchemexpress LLC Kasugamycin | 6980-18-3 | 99.8% | 500 MG
SDP
Catalog No. 5000410817
Encompass_Preferred
Kasugamycin (Ksg) is an antibiotic that binds to both the 30S and 70S ribosomes but not isolated 50S subunits. It mimics mRNA nucleotides to destabilize tRNA binding and inhibit canonical translation initiation.
  • Binds to both 30S and 70S ribosomes
  • Mimics mRNA nucleotides
  • Destabilizes tRNA binding
  • Inhibits canonical translation initiation
Catalog No. 50-004-10817 Supplier Medchemexpress LLC Supplier No. HYB1864500MG
Kasugamycin (Ksg) is an antibiotic that binds to both the 30S and 70S ribosomes but not isolated 50S subunits. It mimics mRNA nucleotides to destabilize tRNA binding and inhibit canonical translation initiation.
  • Binds to both 30S and 70S ribosomes
  • Mimics mRNA nucleotides
  • Destabilizes tRNA binding
  • Inhibits canonical translation initiation