Learn More
Apexbio Technology LLC KASUGAMYCIN-HYDROCHLORIDE-10G
SDP
Catalog No. 5000570371
Encompass_Preferred
Kasugamycin (hydrochloride) (CAS No 19408-46-9) is an aminoglycoside antibiotic widely used in biomedical research primarily for its antibacterial properties Isolated from Streptomyces species it interferes with protein synthesis by inhibiting the translation process on the 30S ribosomal subunit effectively blocking mRNA binding and translation This mechanism makes it a valuable tool for studying bacterial protein synthesis pathways In vitro Kasugamycin demonstrates activity in the nanomolar to low micromolar range making it suitable for precise experimental studies It is commonly utilized in various cell models and animal studies to investigate its effects on bacterial infections as well as in drug screening assays to evaluate novel antibacterial agents Experimental concentrations typically depend on the specific research design allowing flexibility for diverse applications in microbiological and pharmaceutical investigations
Catalog No. 50-005-70371 Supplier Apexbio Technology LLC Supplier No. C550310000
Kasugamycin (hydrochloride) (CAS No 19408-46-9) is an aminoglycoside antibiotic widely used in biomedical research primarily for its antibacterial properties Isolated from Streptomyces species it interferes with protein synthesis by inhibiting the translation process on the 30S ribosomal subunit effectively blocking mRNA binding and translation This mechanism makes it a valuable tool for studying bacterial protein synthesis pathways In vitro Kasugamycin demonstrates activity in the nanomolar to low micromolar range making it suitable for precise experimental studies It is commonly utilized in various cell models and animal studies to investigate its effects on bacterial infections as well as in drug screening assays to evaluate novel antibacterial agents Experimental concentrations typically depend on the specific research design allowing flexibility for diverse applications in microbiological and pharmaceutical investigations