Learn More
Medchemexpress LLC Kasugamycin hydrochloride | 19408-46-9 | 100.0% | 1 ML
SDP
Catalog No. 5000409787
Encompass_Preferred
Kasugamycin (Ksg) hydrochloride hydrate is an antibiotic that binds to 30s and 70s ribosomes, but not to the 50s subunit, and exhibits anti-infective activity. It mimics mRNA nucleotides, disrupting tRNA binding and inhibiting canonical translation initiation. It increases the sensitivity of mycobacteria to Rifampicin in vitro and in mouse infection models.
  • Inhibits P-site tRNA binding by destabilizing mRNA.
  • Decreases the mistranslation rates of asparagine-to-aspartate and glutamine-to-glutamate.
  • Reduces mycobacterial rifampicin phenotypic resistance in wild-type and high mistranslating gatA mutant mycobacterial strains.
  • Results in dose-dependent concentration-time profiles in plasma with rapid clearance of M. tuberculosis in infected mice.
  • Increases Rifampicin susceptibility by 30-fold in mouse models.
Catalog No. 50-004-09787 Supplier Medchemexpress LLC Supplier No. HYB1864A10MM/1ML
Kasugamycin (Ksg) hydrochloride hydrate is an antibiotic that binds to 30s and 70s ribosomes, but not to the 50s subunit, and exhibits anti-infective activity. It mimics mRNA nucleotides, disrupting tRNA binding and inhibiting canonical translation initiation. It increases the sensitivity of mycobacteria to Rifampicin in vitro and in mouse infection models.
  • Inhibits P-site tRNA binding by destabilizing mRNA.
  • Decreases the mistranslation rates of asparagine-to-aspartate and glutamine-to-glutamate.
  • Reduces mycobacterial rifampicin phenotypic resistance in wild-type and high mistranslating gatA mutant mycobacterial strains.
  • Results in dose-dependent concentration-time profiles in plasma with rapid clearance of M. tuberculosis in infected mice.
  • Increases Rifampicin susceptibility by 30-fold in mouse models.