Learn More
Medchemexpress LLC Kasugamycin (hydrochloride) | 19408-46-9 | 99.95% | 25 MG
SDP
Catalog No. 5000413921
Encompass_Preferred
Kasugamycin hydrochloride hydrate is an antibiotic that binds to 30s and 70s ribosomes (but not to the 50s subunit), and exhibits anti-infective activity. It mimics mRNA nucleotides, disrupting tRNA binding and inhibiting canonical translation initiation. It also increases the sensitivity of mycobacteria to Rifampicin in vitro and in mouse infection models.
  • Antibiotic that binds to 30s and 70s ribosomes
  • Mimics mRNA nucleotides
  • Disrupts tRNA binding
  • Inhibits canonical translation initiation
  • Increases sensitivity of mycobacteria to Rifampicin
  • Solid, off-white to light yellow appearance
  • Anti-infective activity
  • Potentiates Rifampicin and limits emergence of resistance in *Mycobacterium tuberculosis*
Catalog No. 50-004-13921 Supplier Medchemexpress LLC Supplier No. 25MGHYB1864A
Kasugamycin hydrochloride hydrate is an antibiotic that binds to 30s and 70s ribosomes (but not to the 50s subunit), and exhibits anti-infective activity. It mimics mRNA nucleotides, disrupting tRNA binding and inhibiting canonical translation initiation. It also increases the sensitivity of mycobacteria to Rifampicin in vitro and in mouse infection models.
  • Antibiotic that binds to 30s and 70s ribosomes
  • Mimics mRNA nucleotides
  • Disrupts tRNA binding
  • Inhibits canonical translation initiation
  • Increases sensitivity of mycobacteria to Rifampicin
  • Solid, off-white to light yellow appearance
  • Anti-infective activity
  • Potentiates Rifampicin and limits emergence of resistance in *Mycobacterium tuberculosis*