Learn More
Selleck Chemical LLC Kasugamycin hydrochloride
SDP
Catalog No. 5000003153
Encompass_Preferred
Kasugamycin is an aminoglycosidic antibiotic isolated from Streptomyces kasugaensis showing broad-spectrum anti-fungal activity against yeast and various fungus species
Catalog No. 50-000-03153 Supplier Selleck Chemical LLC Supplier No. S55531MG
Kasugamycin is an aminoglycosidic antibiotic isolated from Streptomyces kasugaensis showing broad-spectrum anti-fungal activity against yeast and various fungus species