Learn More
Medchemexpress LLC Kasugamycin hydrochloride hydrate | 200132-83-8 | 99.9% | 1 ML
SDP
Catalog No. 5000411392
Encompass_Preferred
Kasugamycin (Ksg) hydrochloride hydrate is an antibiotic that binds to 30s and 70s ribosomes, exhibiting anti-infective activity. It disrupts tRNA binding by mimicking mRNA nucleotides, which inhibits canonical translation initiation. This compound also increases the sensitivity of mycobacteria to Rifampicin both in vitro and in mouse infection models.
  • Antibiotic with anti-infective activity
  • Binds to 30s and 70s ribosomes
  • Mimics mRNA nucleotides, disrupting tRNA binding
  • Inhibits canonical translation initiation
  • Increases sensitivity of mycobacteria to Rifampicin
Catalog No. 50-004-11392 Supplier Medchemexpress LLC Supplier No. HYB1864B10MM/1ML
Kasugamycin (Ksg) hydrochloride hydrate is an antibiotic that binds to 30s and 70s ribosomes, exhibiting anti-infective activity. It disrupts tRNA binding by mimicking mRNA nucleotides, which inhibits canonical translation initiation. This compound also increases the sensitivity of mycobacteria to Rifampicin both in vitro and in mouse infection models.
  • Antibiotic with anti-infective activity
  • Binds to 30s and 70s ribosomes
  • Mimics mRNA nucleotides, disrupting tRNA binding
  • Inhibits canonical translation initiation
  • Increases sensitivity of mycobacteria to Rifampicin