Learn More
Medchemexpress LLC Kasugamycin (hydrochloride hydrate) | 200132-83-8 | 99.9% | 500 MG
SDP
Catalog No. 5000410479
Encompass_Preferred
Kasugamycin (Ksg) hydrochloride hydrate is an antibiotic that binds to 30s and 70s ribosomes but not to the 50s subunit, and has anti-infective activity. It mimics mRNA nucleotides, disrupts tRNA binding and inhibits canonical translation initiation. This product also increases the sensitivity of mycobacteria to Rifampicin in vitro and in mouse infection models.
  • Antibiotic
  • Binds to 30s and 70s ribosomes
  • Anti-infective activity
  • Mimics mRNA nucleotides
  • Disrupts tRNA binding
  • Inhibits canonical translation initiation
  • Increases sensitivity of mycobacteria to Rifampicin
Catalog No. 50-004-10479 Supplier Medchemexpress LLC Supplier No. HYB1864B500MG
Kasugamycin (Ksg) hydrochloride hydrate is an antibiotic that binds to 30s and 70s ribosomes but not to the 50s subunit, and has anti-infective activity. It mimics mRNA nucleotides, disrupts tRNA binding and inhibits canonical translation initiation. This product also increases the sensitivity of mycobacteria to Rifampicin in vitro and in mouse infection models.
  • Antibiotic
  • Binds to 30s and 70s ribosomes
  • Anti-infective activity
  • Mimics mRNA nucleotides
  • Disrupts tRNA binding
  • Inhibits canonical translation initiation
  • Increases sensitivity of mycobacteria to Rifampicin