Learn More
Medchemexpress LLC Kasugamycin hydrochloride (Ksg hydrochloride) | 19408-46-9 | 99.9% | 50 MG
SDP
Catalog No. 5000409904
Encompass_Preferred
Kasugamycin (Ksg) hydrochloride hydrate is an antibiotic that exhibits anti-infective activity by binding to 30s and 70s ribosomes, without affecting the 50s subunit. Its mechanism involves mimicking mRNA nucleotides, which consequently disrupts tRNA binding and hinders canonical translation initiation. This compound has also been observed to enhance the sensitivity of mycobacteria to Rifampicin in both in vitro settings and mouse infection models.
  • Inhibits P-site tRNA binding by destabilizing mRNA.
  • Decreases mistranslation rates of asparagine-to-aspartate and glutamine-to-glutamate.
  • Reduces mycobacterial rifampicin phenotypic resistance in wild-type and high mistranslating gatA mutant mycobacterial strains.
  • Leads to rapid clearance of *M. tuberculosis* in lung burden in Mtb-infected mice.
  • Increases Rifampicin susceptibility by 30-fold.
Catalog No. 50-004-09904 Supplier Medchemexpress LLC Supplier No. HYB1864A50MG
Kasugamycin (Ksg) hydrochloride hydrate is an antibiotic that exhibits anti-infective activity by binding to 30s and 70s ribosomes, without affecting the 50s subunit. Its mechanism involves mimicking mRNA nucleotides, which consequently disrupts tRNA binding and hinders canonical translation initiation. This compound has also been observed to enhance the sensitivity of mycobacteria to Rifampicin in both in vitro settings and mouse infection models.
  • Inhibits P-site tRNA binding by destabilizing mRNA.
  • Decreases mistranslation rates of asparagine-to-aspartate and glutamine-to-glutamate.
  • Reduces mycobacterial rifampicin phenotypic resistance in wild-type and high mistranslating gatA mutant mycobacterial strains.
  • Leads to rapid clearance of *M. tuberculosis* in lung burden in Mtb-infected mice.
  • Increases Rifampicin susceptibility by 30-fold.