Learn More
Cayman Chemical Kasugamycin (hydrochloride), Purity: ≥70%, CAS: 19408-46-9, 100 mg
SDP
Catalog No. 502970789
Encompass_Preferred
An aminoglycoside with diverse biological activities; active against S. dysenteriae, S. flexneri, S. sonnei, and S. equi when at 50 mg/ml and B. melitensis at 6.25 mg/ml; reduces the severity of R. solani-induced rice sheath blight and C. janseana-induced rice narrow brown leaf spot, as well as increases rice yield, at 1 kg/ha; inhibits CHIA (IC50 = 4.13 µM); decreases lung collagen levels in a bleomycin-induced mouse model of pulmonary fibrosis at 12.5, 25, 50, or 100 mg/kg every other day
Catalog No. 50-297-0789 Supplier Cayman Chemical Supplier No. 15322100 MG
An aminoglycoside with diverse biological activities; active against S. dysenteriae, S. flexneri, S. sonnei, and S. equi when at 50 mg/ml and B. melitensis at 6.25 mg/ml; reduces the severity of R. solani-induced rice sheath blight and C. janseana-induced rice narrow brown leaf spot, as well as increases rice yield, at 1 kg/ha; inhibits CHIA (IC50 = 4.13 µM); decreases lung collagen levels in a bleomycin-induced mouse model of pulmonary fibrosis at 12.5, 25, 50, or 100 mg/kg every other day