Learn More
ATCC Kasumi-1; Leukemia (AML); Human (Homo sapiens)
Catalog No. 502384096
Encompass
Kasumi-1 is a myeloblast cell that was isolated from the peripheral blood of an acute myeloblastic leukemia Asian male patient. This cell line was deposited by H Asou in 1989.
Catalog No. 50-238-4096 Supplier ATCC Supplier No. CRL2724
Kasumi-1 is a myeloblast cell that was isolated from the peripheral blood of an acute myeloblastic leukemia Asian male patient. This cell line was deposited by H Asou in 1989.