Learn More
ATCC Kasumi-6; Leukemia; Human (Homo sapiens)
Catalog No. 502384344
Encompass
Kasumi-6 is a myeloblast cell line that was isolated in 1999 from the peripheral blood of an Asian, 64-year-old, male patient with relapsed acute myeloid leukemia, subtype M2. This cell line was deposited by H Asou and can be used in immunology and immune system disorder research.
Catalog No. 50-238-4344 Supplier ATCC Supplier No. CRL2775
Kasumi-6 is a myeloblast cell line that was isolated in 1999 from the peripheral blood of an Asian, 64-year-old, male patient with relapsed acute myeloid leukemia, subtype M2. This cell line was deposited by H Asou and can be used in immunology and immune system disorder research.