Learn More
Med Vet International Kat Kat Adjustable Length Tomcat Catheter, 18cm, J1203BN, Each
Catalog No. 5000581231
Encompass_Preferred
KatKath with KatKlip is an adjustable length catheter with a locking component that lets the clinician adjust the suture ring to desired position 3.5fr up to 18cm length.   Benefits Easy placement Flexible for maximum comfort Thin walled – larger bore lumen Great longitudal strength Rounded atraumatic tip Rotating Suture Collar The KatKath has a rotating suture collar – To allow natural rotation while in situ. More Flexible The catheter is made from a unique non-reactive material that has similar properties to Teflon but is more flexible. More flexibility allows greater comfort. Great Longitudinal Strength The KatKath increased flexibility does not diminish its ability in the effective treatment of blocked cats due to its great longitudinal strength and will not concertina even when encountering gross resistance. Crush Resistant The KatKath is almost impossible to crush and is also highly resistant to kinking.
Catalog No. 50-005-81231 Supplier Med Vet International Supplier No. JOR1203BN
KatKath with KatKlip is an adjustable length catheter with a locking component that lets the clinician adjust the suture ring to desired position 3.5fr up to 18cm length.   Benefits Easy placement Flexible for maximum comfort Thin walled – larger bore lumen Great longitudal strength Rounded atraumatic tip Rotating Suture Collar The KatKath has a rotating suture collar – To allow natural rotation while in situ. More Flexible The catheter is made from a unique non-reactive material that has similar properties to Teflon but is more flexible. More flexibility allows greater comfort. Great Longitudinal Strength The KatKath increased flexibility does not diminish its ability in the effective treatment of blocked cats due to its great longitudinal strength and will not concertina even when encountering gross resistance. Crush Resistant The KatKath is almost impossible to crush and is also highly resistant to kinking.