Learn More
Medchemexpress LLC KAT modulator-1 | 1314006-43-3 | C20H36O2 | 5 MG
SDP
Catalog No. 5000376027
Encompass_Preferred
KAT modulator-1 is a compound identified as a KAT modulator that interacts with p300 full-length, but not its catalytic domain. It is provided as a solid and is intended for laboratory chemicals, manufacture of substances, and epigenetics research, including drug screening applications.
  • Targets histone acetyltransferase
  • Used in cancer research
  • Interacts with p300 full-length
  • Suitable for epigenetics research
  • Identified for laboratory use and drug screening
  • Provided as a solid
  • For research use only
Catalog No. 50-003-76027 Supplier Medchemexpress LLC Supplier No. HY1537685MG
KAT modulator-1 is a compound identified as a KAT modulator that interacts with p300 full-length, but not its catalytic domain. It is provided as a solid and is intended for laboratory chemicals, manufacture of substances, and epigenetics research, including drug screening applications.
  • Targets histone acetyltransferase
  • Used in cancer research
  • Interacts with p300 full-length
  • Suitable for epigenetics research
  • Identified for laboratory use and drug screening
  • Provided as a solid
  • For research use only