Learn More
Medchemexpress LLC KAT modulator-1 (Compound 3) | 1314006-43-3 | 98.0% | 308.50 | C20H36O2 | 10 MG
SDP
Catalog No. 5000287580
Encompass_Preferred
KAT modulator-1 is a small-molecule modulator of lysine acetyltransferase (KAT) activity developed for epigenetics research. It has been shown to interact with full-length p300 but not with the isolated catalytic domain, making it a useful probe for studying KAT-protein interactions and regulatory mechanisms in biochemical and cell-based assays. The compound is supplied as a solid with documented storage recommendations and analytical data.
  • Modulates lysine acetyltransferase (KAT) activity.
  • Interacts with full-length p300 but not the catalytic domain.
  • Suitable for biochemical and cell-based epigenetics studies.
  • Supplied as a solid with defined storage stability.
  • Molecular formula C20H36O2 and molecular weight 308.50.
  • High chemical purity typically reported (about 98%).
Catalog No. 50-002-87580 Supplier Medchemexpress LLC Supplier No. HY15376810MG
KAT modulator-1 is a small-molecule modulator of lysine acetyltransferase (KAT) activity developed for epigenetics research. It has been shown to interact with full-length p300 but not with the isolated catalytic domain, making it a useful probe for studying KAT-protein interactions and regulatory mechanisms in biochemical and cell-based assays. The compound is supplied as a solid with documented storage recommendations and analytical data.
  • Modulates lysine acetyltransferase (KAT) activity.
  • Interacts with full-length p300 but not the catalytic domain.
  • Suitable for biochemical and cell-based epigenetics studies.
  • Supplied as a solid with defined storage stability.
  • Molecular formula C20H36O2 and molecular weight 308.50.
  • High chemical purity typically reported (about 98%).