Learn More
Eurofins Discoverx KAT2A (GCN5L2), 50 µg. (EPI024)
Supplier:  Eurofins Discoverx EPI024
Encompass
KAT2A (GCN5L2) is purified, untagged recombinant KAT2A enzyme. This enzyme contains amino acids 20-341. It retains its binding/enzymatic activity and is intended for use in epigenetic assays. This enzyme is provided as 1 mg/mL frozen in 50% glycerol. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-200-5206