Learn More
Aviva Systems Biology Corp KAT5 Antibody - C-terminal region (ARP38792_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38792_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KAT5. It was validated on Western Blot. . Alternate names: ESA1, HTATIP, HTATIP1, PLIP, TIP, TIP60, cPLA2ApplicationWB, IHC. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-759