Learn More
Aviva Systems Biology Corp Kat5 Antibody - N-terminal region (ARP40066_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40066_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Kat5. It was validated on Western Blot. . Alternate names: 60kDa, AI839539, Htatip, Htatip1, MGC117972, PLIP, Tip55, Tip60, CPLA2ApplicationWB. Species reactivity: Dog, Human, Mouse, Rat
Catalog No. 50-284-850