Learn More
Medchemexpress LLC KAT6A protein, human (biotinylated, Avi) | 93.5% | 10UG
SDP
Catalog No. 5000174622
Encompass_Preferred
KAT6A Protein, Human (Biotinylated, Avi) is a recombinant fragment of the human histone acetyltransferase KAT6A (residues G497-I780), expressed in E. coli and N-terminally labeled with an Avi biotinylation tag. The material is provided at microgram scale for research use, characterized by reducing SDS-PAGE, and has an expected molecular weight of approximately 35.6 kDa.
  • Biotinylated via Avi tag for streptavidin-based capture and detection.
  • Suitable for protein-protein interaction and binding assays.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Recombinantly expressed in E. coli for scalable production.
  • Covers the KAT6A domain G497-I780 for domain-specific studies.
  • Supplied in microgram quantities for biochemical and biophysical assays.
Catalog No. 50-001-74622 Supplier Medchemexpress LLC Supplier No. HYP70323410UG
KAT6A Protein, Human (Biotinylated, Avi) is a recombinant fragment of the human histone acetyltransferase KAT6A (residues G497-I780), expressed in E. coli and N-terminally labeled with an Avi biotinylation tag. The material is provided at microgram scale for research use, characterized by reducing SDS-PAGE, and has an expected molecular weight of approximately 35.6 kDa.
  • Biotinylated via Avi tag for streptavidin-based capture and detection.
  • Suitable for protein-protein interaction and binding assays.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Recombinantly expressed in E. coli for scalable production.
  • Covers the KAT6A domain G497-I780 for domain-specific studies.
  • Supplied in microgram quantities for biochemical and biophysical assays.