Learn More
Medchemexpress LLC KAT7 protein, human (active, N-GST) | >90.0% | 10UG
SDP
Catalog No. 5000174332
Encompass_Preferred
Recombinant human KAT7 protein (active) is produced in Sf9 insect cells with an N-terminal GST tag and supplied as a stabilized, filtered solution for biochemical and enzymatic studies. The preparation is concentrated and quality-tested to support enzyme kinetics, inhibitor screening, and functional assays, with recommended frozen storage to preserve activity.
  • Active enzymatic preparation suitable for biochemical assays.
  • Expressed in Sf9 insect cells for eukaryotic folding.
  • N-terminal GST tag for affinity capture and detection.
  • High purity (>90%) by reducing SDS-PAGE.
  • Supplied in a stabilizing buffer with glycerol and reducing agents.
  • Recommended frozen storage and aliquoting to maintain activity.
  • Available in a small, assay-friendly size.
Catalog No. 50-001-74332 Supplier Medchemexpress LLC Supplier No. HYP70403710UG
Recombinant human KAT7 protein (active) is produced in Sf9 insect cells with an N-terminal GST tag and supplied as a stabilized, filtered solution for biochemical and enzymatic studies. The preparation is concentrated and quality-tested to support enzyme kinetics, inhibitor screening, and functional assays, with recommended frozen storage to preserve activity.
  • Active enzymatic preparation suitable for biochemical assays.
  • Expressed in Sf9 insect cells for eukaryotic folding.
  • N-terminal GST tag for affinity capture and detection.
  • High purity (>90%) by reducing SDS-PAGE.
  • Supplied in a stabilizing buffer with glycerol and reducing agents.
  • Recommended frozen storage and aliquoting to maintain activity.
  • Available in a small, assay-friendly size.