Learn More
CPC Scientific H-Asp-Met-Ser-Ser-Asp-Leu-Glu-Arg-Asp-His-Arg-Pro-His-Val-Ser-Met-Pro-Gln-Asn-Ala-Asn-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific OSTP017B
Encompass
Human calcitonin precursor peptide, called C-procalcitonin, katacalcin or PDN-21, flanking calcitonin on the C-terminal side within the human calcitonin precursor that significantly lowers plasma calcium level.SEQUENCE: H-Asp-Met-Ser-Ser-Asp-Leu-Glu-Arg-Asp-His-Arg-Pro-His-Val-Ser-Met-Pro-Gln-Asn-Ala-Asn-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: DMSSDLERDHRPHVSMPQNANMOLECULAR FORMULA: C97H154N34O36S2MOLECULAR WEIGHT: 2436.6STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [85916-47-8]SYNONYMS: RESEARCH AREA: OsteoporosisREFERENCES:I. Maclntyre et al., Nature, 300, 460 (1982)SKU(s): OSTP-017A, OSTP-017B
Catalog No. 50-211-0030