Learn More
Medchemexpress LLC Katacalcin (TFA) | 99.6% | 2550.62 | 100 MG
SDP
Catalog No. 5000426396
Encompass_Preferred
Katacalcin TFA (PDN 21 TFA) is a potent plasma calcium-lowering peptide from the calcitonin family. It induces a rapid, short-lived decrease in blood calcium and phosphate by promoting bone incorporation. This peptide, encoded by the calc-1 gene alongside calcitonin and procalcitonin NH2-terminal cleavage peptide, effectively inhibits fMLP-induced chemotaxis and deactivates CD14+ peripheral blood mononuclear cell chemotaxis toward various attractants.
  • Potent plasma calcium-lowering peptide
  • Belongs to the calcitonin family
  • Decreases blood calcium and phosphate levels
  • Promotes incorporation into bones
  • Inhibits fMLP-induced chemotaxis
  • Deactivates CD14+ peripheral blood mononuclear cell chemotaxis
  • Regulates CD14+ PBMC migration via protein kinase A-dependent cAMP pathways
Catalog No. 50-004-26396 Supplier Medchemexpress LLC Supplier No. HYP0149A100MG
Katacalcin TFA (PDN 21 TFA) is a potent plasma calcium-lowering peptide from the calcitonin family. It induces a rapid, short-lived decrease in blood calcium and phosphate by promoting bone incorporation. This peptide, encoded by the calc-1 gene alongside calcitonin and procalcitonin NH2-terminal cleavage peptide, effectively inhibits fMLP-induced chemotaxis and deactivates CD14+ peripheral blood mononuclear cell chemotaxis toward various attractants.
  • Potent plasma calcium-lowering peptide
  • Belongs to the calcitonin family
  • Decreases blood calcium and phosphate levels
  • Promotes incorporation into bones
  • Inhibits fMLP-induced chemotaxis
  • Deactivates CD14+ peripheral blood mononuclear cell chemotaxis
  • Regulates CD14+ PBMC migration via protein kinase A-dependent cAMP pathways