Learn More
Medchemexpress LLC Katacalcin TFA | 99.6% | 2550.62 | 25 MG
SDP
Catalog No. 5000425915
Encompass_Preferred
Katacalcin TFA (PDN 21 TFA) is a potent plasma calcium-lowering peptide from the calcitonin family. It rapidly decreases blood calcium and phosphate by promoting their incorporation into bones. It also deactivates CD14+ peripheral blood mononuclear cell (PBMC) chemotaxis and regulates their migration.
  • Potent plasma calcium-lowering peptide
  • Belongs to the calcitonin family
  • Promotes incorporation of calcium and phosphate into bones
  • More potent inhibitor of fMLP-induced chemotaxis
  • Deactivates CD14+ peripheral blood mononuclear cell (PBMC) chemotaxis
  • Regulates human CD14+ PBMC migration
Catalog No. 50-004-25915 Supplier Medchemexpress LLC Supplier No. 25MGHYP0149A
Katacalcin TFA (PDN 21 TFA) is a potent plasma calcium-lowering peptide from the calcitonin family. It rapidly decreases blood calcium and phosphate by promoting their incorporation into bones. It also deactivates CD14+ peripheral blood mononuclear cell (PBMC) chemotaxis and regulates their migration.
  • Potent plasma calcium-lowering peptide
  • Belongs to the calcitonin family
  • Promotes incorporation of calcium and phosphate into bones
  • More potent inhibitor of fMLP-induced chemotaxis
  • Deactivates CD14+ peripheral blood mononuclear cell (PBMC) chemotaxis
  • Regulates human CD14+ PBMC migration