Learn More
World Precision Instrument Katena-Vannas Spring Scissors, 11cm, Straight, 7mm Blades
SDP
Catalog No. 502539424
Encompass
Katena Vannas spring scissors are microsurgical spring scissors used in delicate ophthalmic procedures and other microsurgical applications in a research laboratory. They are named after the renowned Swiss ophthalmic surgeon, Nils Yngve Vannas. Spring scissors can be used ambidextrously, making them favored choice for left-handed people. These scissors are known for their high-quality construction and extreme precision, making them valuable tools in eye surgery. Katena Vannas spring scissors have an elongated, slender design with extra-fine, sharp blades that taper to very fine points. This design allows for intricate and precise cutting in microsurgical settings. They are crafted from high-quality surgical grade stainless steel, ensuring durability, resistance to corrosion, and easy sterilization, meeting the stringent requirements for sterile surgical instruments. These vannas spring scissors are 11 cm (4.3”) long with your choice of tips, 7mm or 10 mm long.
Catalog No. 50-253-9424 Supplier World Precision Instrument Supplier No. 504024
Katena Vannas spring scissors are microsurgical spring scissors used in delicate ophthalmic procedures and other microsurgical applications in a research laboratory. They are named after the renowned Swiss ophthalmic surgeon, Nils Yngve Vannas. Spring scissors can be used ambidextrously, making them favored choice for left-handed people. These scissors are known for their high-quality construction and extreme precision, making them valuable tools in eye surgery. Katena Vannas spring scissors have an elongated, slender design with extra-fine, sharp blades that taper to very fine points. This design allows for intricate and precise cutting in microsurgical settings. They are crafted from high-quality surgical grade stainless steel, ensuring durability, resistance to corrosion, and easy sterilization, meeting the stringent requirements for sterile surgical instruments. These vannas spring scissors are 11 cm (4.3”) long with your choice of tips, 7mm or 10 mm long.