Learn More
Medchemexpress LLC Kathon 886 MW | 55965-84-9 | MFCD01716911 | 14.5% | 264.75 g·mol⁻¹ | C8H9ClN2O2S2 | 1 KG
SDP
Catalog No. 5000299898
Encompass_Preferred
Kathon 886 MW is an aqueous biocide supplied as a 14.5% solution that provides rapid and long-lasting control of microbial contamination in metalworking fluids and metal finishing systems. It is formulated to control bacteria and fungi that cause spoilage, odors, and performance loss, enabling extended fluid life and reduced maintenance costs.
  • Broad-spectrum antimicrobial activity against bacteria and fungi.
  • Effective in soluble, semi-synthetic, and synthetic metalworking fluids.
  • Supplied as a stable aqueous solution for easy dosing.
  • Helps extend fluid life and reduce microbial spoilage.
  • Available with technical and safety documentation for safe handling and use.
Catalog No. 50-002-99898 Supplier Medchemexpress LLC Supplier No. HYW0179821KG
Kathon 886 MW is an aqueous biocide supplied as a 14.5% solution that provides rapid and long-lasting control of microbial contamination in metalworking fluids and metal finishing systems. It is formulated to control bacteria and fungi that cause spoilage, odors, and performance loss, enabling extended fluid life and reduced maintenance costs.
  • Broad-spectrum antimicrobial activity against bacteria and fungi.
  • Effective in soluble, semi-synthetic, and synthetic metalworking fluids.
  • Supplied as a stable aqueous solution for easy dosing.
  • Helps extend fluid life and reduce microbial spoilage.
  • Available with technical and safety documentation for safe handling and use.