Learn More
Medchemexpress LLC Kathon 886MW | 55965-84-9 | MFCD01716911 | 14.5% | 264.8 g/mol | C8H9ClN2O2S2 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW017982100G
Encompass_Preferred
An aqueous CMIT/MIT biocide supplied as a 14.5% active solution, used to control bacterial and fungal growth in metalworking fluids and other water-based industrial systems. Formulated for tankside addition, it provides broad-spectrum antimicrobial protection to extend fluid life and reduce microbial spoilage in recirculating systems.
  • Broad-spectrum antimicrobial activity against bacteria and fungi.
  • Aqueous CMIT/MIT formulation at 14.5% active concentration.
  • Suitable for metalworking fluids and other water-based industrial systems.
  • Effective for tankside preservation and microbicide treatment.
  • Helps reduce downtime by extending fluid life and preventing biodegradation.
  • Formaldehyde-free formulation.
Catalog No. 50-003-01649