Learn More
ATCC KATO III; Gastric Carcinoma; Human (Homo sapiens)
Supplier:  ATCC HTB103
Encompass
KATO III cells have spherical morphology and were established in vitro from a pleural effusion of a 55-year-old, Asian male stomach cancer patient. Use these cells in cancer research.
Catalog No. 50-238-1857