Learn More
Cayman Chemical KaurenoIc AcId 10mg
SDP
Catalog No. 502938991
Encompass_Preferred
A diterpene with diverse biological activities; cytotoxic to a variety of prostate, colon, and breast cancer cells (IC50s = 15.03-48.87 μg/ml); active against S. epidermidis, S. capitis, E. faecalis, and S. haemolyticus in vitro (MICs = 6.25-12.5 μg/ml); reduces colonic MPO activity and MDA levels in a rat model of acetic acid-induced colitis at 50 and 100 mg/kg, as well as colonic tissue damage, leukocytic infiltration, and submucosal edema when administered at a dose of 100 mg/kg
Catalog No. 50-293-8991 Supplier Cayman Chemical Supplier No. 2895710 MG
A diterpene with diverse biological activities; cytotoxic to a variety of prostate, colon, and breast cancer cells (IC50s = 15.03-48.87 μg/ml); active against S. epidermidis, S. capitis, E. faecalis, and S. haemolyticus in vitro (MICs = 6.25-12.5 μg/ml); reduces colonic MPO activity and MDA levels in a rat model of acetic acid-induced colitis at 50 and 100 mg/kg, as well as colonic tissue damage, leukocytic infiltration, and submucosal edema when administered at a dose of 100 mg/kg