Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kauri Gum Solution (In Butanol), Fisher Chemical

Catalog No. SK71QT Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 qt.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SK7-1QT 1 qt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SK7-1QT
  • $1,265.00
    $1,018.65 / Each

    Save $246.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $11,670.00
    $9,401.65 / Case of 12 EA

    Save $2,268.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemical Name or Material Kauri Gum Solution
Color Yellow
Name Note In Butanol
CAS 77465-88-4, 71-36-3
Boiling Point 117.6°C
Melting Point -89°C
Molecular Formula C4H10O
MDL Number MFCD00212241
Density 0.810g/cm³
Packaging Metal Can
Viscosity >3 mPa/s at 20°C
Vapor Pressure 6.7mbar at 20°C
Quantity 1 qt.
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.