Learn More
Enzo Life Sciences Kavain (+/-) (50mg). CAS: 500-64-1
SDP
Catalog No. 502011180
Encompass_Preferred
Kavain is one constituent of kava-kava, an herbal drug of the South Pacific islands, which displays anti-convulsive, anxiolytic, analgesic and muscle relaxing properties. Kavain inhibits voltage-dependent Na+ channels in rat CA1 hippocampal neurons. It displays protective effects in the MPTP mouse model of Parkinson's disease but potentiates NMDA toxicity in the developing hippocampus. Alternative name: (+/-)-4-Methoxy-6-styryl-5 6-dihydro-alpha-pyrone. Purity: ≥98%. Long Term Storage: Ambient.
Catalog No. 50-201-1180 Supplier Enzo Life Sciences Supplier No. BMLNA1360050
Kavain is one constituent of kava-kava, an herbal drug of the South Pacific islands, which displays anti-convulsive, anxiolytic, analgesic and muscle relaxing properties. Kavain inhibits voltage-dependent Na+ channels in rat CA1 hippocampal neurons. It displays protective effects in the MPTP mouse model of Parkinson's disease but potentiates NMDA toxicity in the developing hippocampus. Alternative name: (+/-)-4-Methoxy-6-styryl-5 6-dihydro-alpha-pyrone. Purity: ≥98%. Long Term Storage: Ambient.