Learn More
Medchemexpress LLC (+)-Kavain | 500-64-1 | 99.1% | 5 MG
SDP
Catalog No. 501932987
Encompass_Preferred
(+)-Kavain is a primary kavalactone derived from Piper methysticum, known for its anticonvulsive properties. It functions by interacting with voltage-dependent Na+ and Ca2+ channels, thereby attenuating vascular smooth muscle contraction. Additionally, it has been shown to bind to the α4β2δ GABAA receptor, enhancing GABA efficacy. Its anti-inflammatory actions have been extensively researched, making it a subject of study for inflammatory disease treatments.
  • Attenuates vascular smooth muscle contraction by interacting with voltage-dependent Na+ and Ca2+ channels.
  • Potentiates GABAA receptors by binding to the α4β2δ subtype.
  • Exhibits anti-inflammatory effects.
Catalog No. 50-193-2987 Supplier Medchemexpress LLC Supplier No. HYB16715MG
(+)-Kavain is a primary kavalactone derived from Piper methysticum, known for its anticonvulsive properties. It functions by interacting with voltage-dependent Na+ and Ca2+ channels, thereby attenuating vascular smooth muscle contraction. Additionally, it has been shown to bind to the α4β2δ GABAA receptor, enhancing GABA efficacy. Its anti-inflammatory actions have been extensively researched, making it a subject of study for inflammatory disease treatments.
  • Attenuates vascular smooth muscle contraction by interacting with voltage-dependent Na+ and Ca2+ channels.
  • Potentiates GABAA receptors by binding to the α4β2δ subtype.
  • Exhibits anti-inflammatory effects.