Learn More
Medchemexpress LLC KB-0742 dihydrochloride | 2416874-75-2 | 98.8% | 360.33 | 10 MG
SDP
Catalog No. 5000365644
Encompass_Preferred
KB-0742 dihydrochloride is a potent, selective, and orally active CDK9 inhibitor with an IC50 of 6 nM for CDK9/cyclin T1. It exhibits significant selectivity over other CDK kinases and demonstrates potent anti-tumor activity.
  • Potent, selective, and orally active CDK9 inhibitor
  • Shows over 50-fold selectivity for CDK9/cyclin T1
  • Exhibits potent anti-tumor activity
  • Significantly reduces downstream phosphorylation of RNA Pol II
  • Reduces global androgen receptor protein levels
  • Shows cytostatic effects in prostate cancer and leukemia cell lines
  • Well tolerated and reduces tumor burden in xenograft models
  • Soluble in H2O and DMSO
Catalog No. 50-003-65644 Supplier Medchemexpress LLC Supplier No. 10MGHY137478A
KB-0742 dihydrochloride is a potent, selective, and orally active CDK9 inhibitor with an IC50 of 6 nM for CDK9/cyclin T1. It exhibits significant selectivity over other CDK kinases and demonstrates potent anti-tumor activity.
  • Potent, selective, and orally active CDK9 inhibitor
  • Shows over 50-fold selectivity for CDK9/cyclin T1
  • Exhibits potent anti-tumor activity
  • Significantly reduces downstream phosphorylation of RNA Pol II
  • Reduces global androgen receptor protein levels
  • Shows cytostatic effects in prostate cancer and leukemia cell lines
  • Well tolerated and reduces tumor burden in xenograft models
  • Soluble in H2O and DMSO