Learn More
Medchemexpress LLC HY-137478A 5mg Medchemexpress, KB-0742 (dihydrochloride) CAS:2416874-75-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY137478A5MG
Encompass_Preferred
Medchemexpress, HY-137478A 5mg KB-0742 (dihydrochloride) CAS:2416874-75-2 KB-0742 dihydrochloride is a potent, selective and orally active CDK9 inhibitor with an IC50 of 6 nM for CDK9/cyclin T1. KB-0742 dihydrochloride is selective for CDK9/cyclin T1 with >50-fold selectivity over other CDK kinases. KB-0742 dihydrochloride has potent anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1492